Learn More
Meso Scale Discovery S-PLEX Human IL-10 KitQP
SDP
Supplier:  Meso Scale Discovery K151Y2S21
Encompass
The Meso Scale Discovery S-PLEX Human IL-10 Kit is a multiplex immunoassay that measures IL-10 levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescent detection technology and is run on Meso Scale Discovery instrumentation The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags and calibrator diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL The total assay time is 3 5 hours This product can be used to measure IL-10 levels in cytokine release assays or to profile samples from clinical research studies
Catalog No. 50-000-78781