Learn More
Meso Scale Discovery S-PLEX Human IL-12p70 Kit
SDP
Supplier:  Meso Scale Discovery K151G3S2
Encompass
The Meso Scale Discovery S-PLEX Human IL-12p70 Kit is a multiplex immunoassay that measures human IL-12p70 in serum plasma cell culture supernatants and other biological samples This assay uses electrochemiluminescence (ECL) detection technology and is designed for use on Meso Scale Discovery instrumentation The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag and calibrator solutions to generate a standard curve This assay has high sensitivity broad dynamic range and requires low sample volumes The S-PLEX Human IL-12p70 Kit provides quantitative determination of IL-12p70 levels in approximately 4 hours
Catalog No. 50-000-79653