Learn More
Meso Scale Discovery S-PLEX Human IL-12p70 KitQP
SDP
Supplier:  Meso Scale Discovery K151G3S21
Encompass
The Meso Scale Discovery S-PLEX Human IL-12p70 Kit is an immunoassay for measuring human interleukin 12 (IL-12) in various samples It uses electrochemiluminescence (ECL) detection for high sensitivity and a broad dynamic range with low sample volume The assay is specific for IL-12p70 with minimal cross-reactivity The kit includes a 96-well plate antibodies buffers diluent and calibrators It takes 3 5 hours to complete and provides enough reagents for one 96-well plate
Catalog No. 50-000-78774