Learn More
Meso Scale Discovery S-PLEX Human IL-13 KitQP
SDP
Supplier:  Meso Scale Discovery K151Y9S21
Encompass
The Meso Scale Discovery S-PLEX Human IL-13 Kit is a multiplex immunoassay that allows simultaneous measurement of IL-13 biomarker levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluents optimized for human samples The assay takes approximately 3 hours to complete and is designed for use on Meso Scale Discovery instrumentation
Catalog No. 50-000-79821