Learn More
Meso Scale Discovery S-PLEX Human IL-17A KitQP
SDP
Supplier:  Meso Scale Discovery K151C3S21
Encompass
The Meso Scale Discovery S-PLEX Human IL-17A Kit is a multiplex immunoassay that allows simultaneous measurement of IL-17A in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium read buffer wash buffer blocker solutions calibrator and controls The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-80164