Learn More
Meso Scale Discovery S-PLEX Human IL-2 Kit 25 PL
SDP
Supplier:  Meso Scale Discovery K151Z2S4
Encompass
The Meso Scale Discovery S-PLEX Human IL-2 Kit is a multiplex immunoassay that measures IL-2 in human serum plasma and cell culture supernatant The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and diluent The kit can test up to 80 samples in duplicate and has a testing time of 3 5 hours
Catalog No. 50-000-79865
Add to Cart
Request a Quote