Learn More
Meso Scale Discovery S-PLEX Human IL-2 KitQP
SDP
Supplier:  Meso Scale Discovery K151Z2S21
Encompass
The Meso Scale Discovery S-PLEX Human IL-2 Kit is a multiplex immunoassay that measures human interleukin-2 (IL-2) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and diluent The assay takes approximately 3 5 hours to complete and has a detection range from 0 06-2500 pg/mL
Catalog No. 50-000-79242