Learn More
Meso Scale Discovery S-PLEX Human IL-21 KitQP
SDP
Supplier:  Meso Scale Discovery K151ADNS21
Encompass
The S-PLEX Human IL-21 Kit from Meso Scale Discovery is an immunoassay kit for the quantitative measurement of human interleukin 21 (IL-21) in serum plasma and cell culture supernatants The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label assay diluent calibrator and wash buffer The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL
Catalog No. 50-000-80359