Learn More
Meso Scale Discovery S-PLEX Human IL-22 Kit
SDP
Supplier:  Meso Scale Discovery K151H3S1
Encompass
The Meso Scale Discovery S-PLEX Human IL-22 Kit is an immunoassay that enables the simultaneous detection and quantification of the human cytokine interleukin-22 (IL-22) in cell culture supernatants serum plasma and other biological matrices This multiplex kit utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrator diluent The assay has a typical lower limit of detection of 0 04 pg/mL and an upper limit of quantification of 10 000 pg/mL for IL-22
Catalog No. 50-000-79695