Learn More
Meso Scale Discovery S-PLEX Human IL-22 KitQP
SDP
Supplier:  Meso Scale Discovery K151H3S22
Encompass
The Meso Scale Discovery S-PLEX Human IL-22 Kit is a multiplex immunoassay that measures IL-22 in human serum plasma and cell culture supernatant The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium and calibrators and controls needed to quantify IL-22 levels It can accurately quantify IL-22 in the range of 0 06-2500 pg/mL The assay takes approximately 3 5 hours to complete This kit allows for the precise sensitive and rapid measurement of IL-22
Catalog No. 50-000-82333