Learn More
Meso Scale Discovery S-PLEX Human IL-4 KitQP
SDP
Supplier:  Meso Scale Discovery K151A3S21
Encompass
The S-PLEX Human IL-4 Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of human interleukin 4 (IL-4) in serum plasma and cell culture supernatants The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label calibrators diluent read buffer wash buffer and plate seals The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-79618