Learn More
Meso Scale Discovery S-PLEX Human IL-4 KitQP
SDP
Supplier:  Meso Scale Discovery K151A3S22
Encompass
The Meso Scale Discovery S-PLEX Human IL-4 Kit is an immunoassay that enables the simultaneous detection of human interleukin 4 (IL-4) using electrochemiluminescence (ECL) technology on the MSD platform This multiplex assay kit includes a plate coated with capture antibodies detection antibodies labeled with an ECL tag and calibrators and controls to generate a standard curve and validate assay performance The assay has a sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for human IL-4 This kit allows for the quantitative measurement of IL-4 levels in human serum plasma and cell culture supernatants
Catalog No. 50-000-80388