Learn More
Meso Scale Discovery S-PLEX Human IL-5 Kit
SDP
Supplier:  Meso Scale Discovery K151J3S1
Encompass
The Meso Scale Discovery S-PLEX Human IL-5 Kit is a multiplex immunoassay for measuring IL-5 in human serum plasma and cell culture supernatant It includes a 96-well plate with capture and detection antibodies Read Buffer Diluent 2 Calibrator Blend and sample diluent The assay detects 0 03-2500 pg/mL using 50 L of sample employing electroluminescent technology for high sensitivity and a wide dynamic range It profiles samples for biomarker research in asthma allergies and parasitic infections
Catalog No. 50-000-82477