Learn More
Meso Scale Discovery S-PLEX Human Tau PT181 Kit, 5 plate size, 96-well plate, LLOD 77 fg/mL, dynamic range 77 - 990,000 fg/mL, human species reactivity, mouse monoclonal antibodies
SDP
Supplier:  Meso Scale Discovery K151AGMS2
Encompass
The MESO SCALE DISCOVERY S-PLEX Human Tau (pT181) Kit is an advanced immunoassay designed for measuring phosphorylated Tau (Threonine 181) in human biological samples. It utilizes the S-PLEX platform, which greatly enhances assay sensitivity and improves the ability to detect subtle changes in Tau levels, vital for neurobiology, neurodegeneration, and Alzheimer's disease research. This assay is compatible with various sample types, providing a broad dynamic range and high-specificity detection of Tau levels.
  • Assay platform optimized for ultrasensitivity.
  • Detects phosphorylated Tau (pT181) in human samples.
  • Significantly reduced lower limit of detection.
  • Compatible with serum, plasma, CSF, cell culture supernatant, and urine.
  • Dynamic range of 77 fg/mL to 990,000 fg/mL.
  • Usage limited to research settings only.
Catalog No. NC3739856
Only null left
Add to Cart
Request a Quote