Learn More
Meso Scale Discovery S-PLEX Human Tau (pT217) Kit, 880 fg/mL LLOD, dynamic range 880 - 3,590,000 fg/mL, 96-well plate, mouse monoclonal capture and detection antibody
SDP
Supplier:  Meso Scale Discovery K151APFS2
Encompass
S-PLEX Human Tau (pT217) Kit, 880 fg/mL LLOD, dynamic range 880 - 3,590,000 fg/mL, 96-well plate, mouse monoclonal capture and detection antibody
Catalog No. NC3739862
Add to Cart
Request a Quote