Learn More
Meso Scale Discovery S-PLEX Human Tau pT231 KitQP
SDP
Supplier:  Meso Scale Discovery K151AGNS21
Encompass
The S-PLEX Human Total Tau Kit from Meso Scale Diagnostics is an electrochemiluminescent immunoassay for the quantitative measurement of total tau protein in human serum plasma and CSF The kit contains a 96-well plate coated with a monoclonal antibody to capture total tau a ruthenium-conjugated detection antibody calibrators controls 10X diluent read buffer wash buffer and plate seals The assay has a range of 50-2000 pg/mL and takes 3 5 hours to complete It uses MSD s proprietary S-PLEX technology based on electrochemiluminescence providing high sensitivity and a wide dynamic range The kit includes all necessary reagents to complete a 96-well assay
Catalog No. 50-000-79520