Learn More
Meso Scale Discovery S-PLEX Human TNF-B Kit 25 PL
SDP
Supplier:  Meso Scale Discovery K151ADPS4
Encompass
The Meso Scale Discovery K151ADPS-4 is a 4-plex assay kit for measuring human IL-1 IL-6 IL-8 and TNF- in serum plasma cell culture supernatants and other biological samples The assay uses MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and a 10X read buffer for approximately 40 tests in duplicate The assay has a short protocol with no wash steps required and results can be read on MSD s SECTOR Imager within 3 5 hours
Catalog No. 50-000-79496
Add to Cart
Request a Quote