Learn More
Meso Scale Discovery S-PLEX Human TNF- KitQP
SDP
Supplier:  Meso Scale Discovery K151E3S22
Encompass
The Meso Scale Discovery S-PLEX Human TNF-alpha Kit is a multiplex immunoassay that measures human TNF-alpha in serum plasma and cell culture supernatants The assay uses electrochemiluminescent detection technology and provides high sensitivity and broad dynamic range The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and diluent The assay takes 3 5 hours to complete and requires a Meso Scale Discovery instrument for analysis The kit can test up to 80 samples in duplicate and has a lower limit of detection of 0 04 pg/mL
Catalog No. 50-000-82435