Learn More
Meso Scale Discovery S-PLEX NHP CTLA-4 Kit 25 PL
SDP
Supplier:  Meso Scale Discovery K156AGLS4
Encompass
The Meso Scale Discovery CTLA-4 Discovery Kit is a 96-well plate assay that measures the levels of CTLA-4 in biological samples such as serum plasma cell culture supernatants cell lysates or other biological fluids It uses electrochemiluminescence (ECL) technology and provides high sensitivity broad dynamic range and excellent reproducibility The assay takes approximately 3 5 hours to complete and requires a Meso Scale Discovery SECTOR Imager for readout This product allows for the quantitative measurement of CTLA-4 which plays an important role in downregulating immune responses
Catalog No. 50-000-80872
Only null left
Add to Cart
Request a Quote