Learn More
Meso Scale Discovery S-PLEX NHP CTLA-4 Kit
SDP
Supplier:  Meso Scale Discovery K156AGLS2
Encompass
The Meso Scale Discovery CTLA-4 Discovery Kit is a 96-well plate assay that measures CTLA-4 levels in biological samples such as serum plasma cell culture supernatants and other body fluids The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity a broad dynamic range and requires low sample volumes The kit includes a 96-well plate coated with capture antibody detection antibody read buffer sample diluent and calibrator The total assay time is 3 5 hours This product can aid research on CTLA-4 expression and the effects of CTLA-4-targeting therapeutics
Catalog No. 50-000-78611
Add to Cart
Request a Quote