Learn More
Meso Scale Discovery S-PLEX NHP CTLA-4 KitQP
SDP
Supplier:  Meso Scale Discovery K156AGLS21
Encompass
The Meso Scale Discovery CTLA-4 Discovery Kit is a 96-well immunoassay that quantitatively measures human CTLA-4 protein in serum plasma cell culture supernatants cell and tissue lysates The assay uses MSD electrochemiluminescence detection technology and provides high sensitivity a wide dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer 10X Diluent and calibrators and controls to generate a standard curve The assay takes 3 5 hours total time and has a lower limit of detection of 156 pg/mL CTLA-4
Catalog No. 50-000-81030