Learn More
Meso Scale Discovery S-PLEX NHP IL-2 Kit
SDP
Supplier:  Meso Scale Discovery K156Z2S1
Encompass
The Meso Scale Discovery S-PLEX Human IL-6 Kit is an immunoassay that uses electrochemiluminescence (ECL) technology for the quantitative measurement of human interleukin 6 (IL-6) in serum plasma and cell culture supernatants This assay kit contains a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators diluent and wash buffer The assay has a sensitivity of 0 3 pg/mL and a broad dynamic range from 0 3-10 000 pg/mL This product can be used to measure IL-6 levels for the study of inflammatory diseases and immune system function
Catalog No. 50-000-78493