Learn More
Meso Scale Discovery S-PLEX NHP IL-6 Kit
SDP
Supplier:  Meso Scale Discovery K156B3S1
Encompass
The Meso Scale Discovery S-PLEX Human IL-6 Kit is an immunoassay using electrochemiluminescence (ECL) for measuring IL-6 in human serum plasma and cell culture supernatant It employs SULFO-TAG labels that emit light when stimulated via carbon electrodes The kit includes a 96-well plate with capture antibodies for IL-6 detection antibodies with ruthenium labels calibrators and diluent Sensitivity is 0 3 pg/mL with an upper limit of 10 000 pg/mL It requires 50 L of sample and is compatible with MSD platforms like SECTOR Imager and QuickPlex
Catalog No. 50-000-79312