Learn More
Meso Scale Discovery S-PLEX NHP Neurofilament L Kit
SDP
Supplier:  Meso Scale Discovery K156AKGS2
Encompass
The Meso Scale Discovery Non-Human Primate Neurofilament Light Chain S-Plex Assay Kit measures neurofilament light chain (NfL) in non-human primate samples like serum plasma CSF and brain tissue lysates It uses electrochemiluminescence detection for high sensitivity a wide dynamic range and low sample volume The kit includes a 96-well plate with capture antibody detection antibody with an electrochemiluminescent label calibrators controls buffers blocker solutions and plate seals This assay quantifies NfL a biomarker of axonal injury and neurodegeneration
Catalog No. 50-000-78610
Only null left
Add to Cart
Request a Quote