Learn More
Meso Scale Discovery S-PLEX NHP Tau pT181 Kit
SDP
Supplier:  Meso Scale Discovery K156AGMS2
Encompass
The Meso Scale Discovery Tau (pT181) Kit is an electrochemiluminescent immunoassay designed for the quantitative measurement of total tau phosphorylated at threonine 181 in human serum plasma and CSF The assay uses MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide a highly sensitive and reproducible method for measuring phosphorylated tau levels The kit includes a 96-well plate coated with a tau (pT181) capture antibody a SULFO-TAG labeled detection antibody assay diluent calibrator and wash buffer The assay has a typical lower limit of quantification of 1 95 pg/mL and an upper limit of quantification of 1000 pg/mL
Catalog No. 50-000-80858
Only null left
Add to Cart
Request a Quote