Learn More
Meso Scale Discovery S-PLEX SARS-CoV-2 N Kit
SDP
Supplier:  Meso Scale Discovery K150ADHS1
Encompass
The S-PLEX SARS-CoV-2 N Kit is a multiplex immunoassay panel for the simultaneous qualitative detection of nucleocapsid protein antigens from SARS-CoV-2 in human serum plasma and saliva This assay uses electrochemiluminescence (ECL) on the SECTOR Imager 2400 platform The kit includes a plate coated with capture antibodies detection antibodies conjugated to ruthenium and calibrators and controls needed to run the assay Results are obtained in under 3 hours This assay allows sensitive high-throughput testing for SARS-CoV-2 antigens
Catalog No. 50-000-78814