Learn More
Medchemexpress LLC (S)-Pro-xylane | 868156-46-1 | MFCD32067869 | >99.0% | 192.21 g/mol | C8H16O5 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY108036A10MM 1ML
Encompass_Preferred
A ready-to-use 10 mM solution of (S)-Pro-xylane in DMSO supplied for biochemical and cosmetic research. (S)-Pro-xylane is a bioactive C-glycoside that stimulates glycosaminoglycan biosynthesis in fibroblasts, supports dermal extracellular matrix stability, and promotes collagen and hyaluronic acid production to improve skin hydration and elasticity.
  • Stimulates glycosaminoglycan synthesis in fibroblasts.
  • Promotes collagen and hyaluronic acid production.
  • Improves skin hydration and elasticity in topical models.
  • Supplied as a 10 mM solution in DMSO for immediate use.
  • Eco-friendly and biodegradable profile.
Catalog No. 50-002-43020