Learn More
Abcam (S)-Propranolol hydrochloride, beta adrenoceptor antagonist, 100MG
Supplier:  Abcam AB141133100MG
Encompass_Preferred
MW 295.8 Da, Purity >98%. Non-selective β adrenoceptor antagonist. More active enantiomer of propranolol (ab120757).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6865