Learn More
Cayman Chemical S- Proprnololhydrochl 250mg
SDP
Supplier:  Cayman Chemical 17291250 MG
Encompass_Preferred
The active enantiomer of propranolol, a β-adrenergic receptor antagonist with log Kd values of -8.16, -9.08, and -6.93 for β1, β2, and β3, respectively; widely used in clinical practice for the treatment of hypertension, angina pectoris, and cardiac ischemia
Catalog No. 50-296-5428