Learn More
Medchemexpress LLC (S)-Purvalanol B | >=98.0% | 5 MG
SDP
Supplier:  Medchemexpress LLC HY18299B5MG
Encompass_Preferred
Purvalanol B (NG 95) is a potent, selective, reversible, and ATP-competitive inhibitor of CDK. It inhibits the growth of a chloroquine-resistant strain of P. falciparum.
  • Potent, selective, reversible, and ATP-competitive inhibitor of CDK
  • Inhibits cdc2-cyclin B, CDK2-cyclin A, CDK5-p35, and CDK2-cyclin E
  • Selective for CDK over other protein kinases
  • Inhibits growth of chloroquine-resistant P. falciparum
  • Inhibitory activity against Cyclin-dependent kinase 1-cyclin B complex from starfish oocytes
Catalog No. 50-003-87677