Learn More
Medchemexpress LLC (S)-pyrrolidine-2-carboxamide hydrochloride | 42429-27-6 | MFCD00058156 | 97.0% | 150.61 g/mol | C5H11ClN2O | 1 G

Supplier: Medchemexpress LLC HYW0174131G
(S)-Pyrrolidine-2-carboxamide hydrochloride is the S-enantiomer of a proline-derived carboxamide supplied as the hydrochloride salt for research use. It serves as a chiral building block in medicinal chemistry and biochemical synthesis and is identified by CAS 42429-27-6.
- Chiral proline derivative useful as a synthetic building block.
- Provided as the hydrochloride salt, typically a white to off-white solid.
- Molecular formula C5H11ClN2O; molecular weight 150.61 g/mol.
- Commonly supplied at research-grade purity (for example, ~97% reported by suppliers).
- Storage recommendations: powder at -20°C (shelf life 3 years) or 4°C (shelf life 2 years); in solution keep at -80°C (shelf life 6 months).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.