Learn More
Medchemexpress LLC (S)-pyrrolidine-2-carboxamide hydrochloride | 42429-27-6 | MFCD00058156 | 97.0% | 150.61 g·mol⁻¹ | C5H11ClN2O | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0174135G
Encompass_Preferred
(S)-Pyrrolidine-2-carboxamide hydrochloride is the S-enantiomer of prolinamide supplied as the hydrochloride salt. It appears as a white to off-white solid and is used as an amino-acid derivative building block in synthetic chemistry and life-science research. It is typically handled at laboratory scale for small-molecule synthesis and analytical applications.
  • White to off-white solid form for easy handling.
  • Hydrochloride salt improves stability and solubility in polar solvents.
  • Useful chiral building block for synthesis of proline derivatives and analogs.
  • Suitable for research-scale synthesis and analytical studies.
  • Well-characterized compound with established molecular weight and formula.
Catalog No. 50-002-99953