Learn More
Medchemexpress LLC (S,R)-GSK321 | 1816272-18-0 | 98.6% | 50 MG

Supplier: Medchemexpress LLC HY128888B50MG
(S,R)-GSK321 is the (S,R)-enantiomer of GSK321, a potent and selective mutant IDH1 inhibitor with strong activity against various mutant IDH1 forms and high selectivity over IDH2. It induces a decrease in intracellular 2-HG, helps abrogate myeloid differentiation block, and promotes granulocytic differentiation in leukemic blasts and more immature stem-like cells. This compound is intended for research use in studies related to acute myeloid leukemia (AML) and other cancers, and is not sold to patients.
- Potent and selective mutant IDH1 inhibitor
- High selectivity over IDH2
- Induces a decrease in intracellular 2-HG
- Abrogates myeloid differentiation block
- Induces granulocytic differentiation
- Suitable for research on acute myeloid leukemia (AML) and other cancers
- Molecular weight: 501.55
- Chemical formula: C28H28FN5O3
- Appearance: white to off-white solid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.