Learn More
Medchemexpress LLC (S,R,S)-AHPC | 1448297-52-6 | MFCD29049827 | 99.1% | 430.56 g/mol | C22H30N4O3S | 100MG
SDP
Supplier:  Medchemexpress LLC HY125845100MG
Encompass_Preferred
(S,R,S)-AHPC is a VH032-derived VHL ligand used to recruit the von Hippel-Lindau E3 ligase in PROTAC and targeted protein degradation research. Supplied as a powder for research use, it has CAS 1448297-52-6, chemical formula C22H30N4O3S, molecular weight 430.56 g/mol, and measured HPLC purity 99.13%.
  • Provides high-purity VHL ligand suitable for PROTAC construction.
  • Powder form for easy storage and handling.
  • Stable under recommended storage conditions (powder: -20°C/4°C; in solvent: -80°C/-20°C).
  • Soluble in DMSO and water with ultrasonic assistance for formulation.
  • Available in small research pack sizes for early-stage studies.
  • Characterized by HPLC and LCMS to confirm identity and purity.
Catalog No. 50-002-34920