Learn More
Medchemexpress LLC (S,R,S)-AHPC-3-methylbutanyl acetate-methanesulfonothioate-Me-C10-NH2 TFA | 2417370-89-7 | 95.1% | C41H62F3N5O10S3 | 10MG

Supplier: Medchemexpress LLC HY138551A10MG
This compound is an E3 ligase ligand-linker conjugate incorporating the (S,R,S)-AHPC ligand, provided as a viscous liquid for use in PROTAC synthesis and related chemical biology research. Supplied for research use only.
- Purity 95.1%.
- Molecular formula C41H62F3N5O10S3.
- Molecular weight 938.15 g·mol⁻¹.
- Appearance viscous liquid, colorless to light yellow.
- Solubility in DMSO 100 mg/mL; ultrasonic agitation recommended.
- Storage: 4°C; in solvent -80°C (6 months) or -20°C (1 month); protect from light and store under nitrogen.
- Suitable for PROTAC ligand-linker conjugate synthesis and biochemical studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.