Learn More
Medchemexpress LLC (S,R,S)-AHPC-Boc | 1448189-98-7 | MFCD31692982 | 96.4% | 530.68 | C27H38N4O5S | 100MG
SDP
Supplier:  Medchemexpress LLC HY123109100MG
Encompass_Preferred
Boc-protected AHPC (VH032-Boc) is a small-molecule ligand that recruits the von Hippel-Lindau (VHL) E3 ligase and is commonly used as the VHL-recruiting moiety in PROTAC design and E3 ligase binding studies. It is supplied as a solid reagent for synthetic chemistry and biochemical assays, and is characterized and documented for research use.
  • Ligand for von Hippel-Lindau (VHL) E3 ligase.
  • Intended for use as the recruiting moiety in PROTAC design.
  • Supplied as a solid for organic synthesis and biochemical assays.
  • High purity (≈96.4%) suitable for research applications.
  • Characterized by NMR and LCMS; COA and SDS available.
Catalog No. 50-002-36994