Learn More
Medchemexpress LLC (S,R,S)-AHPC-Boc | 1448189-98-7 | MFCD31692982 | 96.4% | 530.68 g/mol | C27H38N4O5S | 25MG
SDP
Supplier:  Medchemexpress LLC HY12310925MG
Encompass_Preferred
(S,R,S)-AHPC-Boc (VH032-Boc) is a Boc-protected small-molecule ligand that recruits the von Hippel-Lindau (VHL) E3 ubiquitin ligase for use in targeted protein degradation and PROTAC design. It is supplied at research-grade purity for synthetic conjugation and downstream chemical modification in chemical biology and medicinal chemistry workflows.
  • Boc-protected VHL ligand enabling linker attachment for conjugation
  • Suitable for PROTAC synthesis and VHL recruitment studies
  • High purity appropriate for research and synthetic use
  • Available in multiple pack sizes to support small-scale and scale-up work
  • Compatible with common organic synthesis and conjugation chemistries
Catalog No. 50-002-37372