Learn More
Medchemexpress LLC (S,R,S)-AHPC-Boc derivative 1 | 2086298-33-9 | 98.6% | 560.71 | 100 MG

Supplier: Medchemexpress LLC HY173097100MG
This product is a chemical compound primarily intended for laboratory use and the manufacture of substances. It is designed for research purposes only.
- White to off-white solid appearance
- High purity at 98.6% (LCMS)
- Consistent structure confirmed by 1H NMR and LCMS
- Stable for 3 years as a powder at -20°C
- Stable for 6 months in solvent at -80°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.