Learn More
Medchemexpress LLC (S,R,S)-AHPC-Boc derivative 1 | 2086298-33-9 | 98.6% | 560.71 | 50 MG

Supplier: Medchemexpress LLC HY17309750MG
(S,R,S)-AHPC-Boc derivative 1 is a chemical substance intended for laboratory research and the manufacture of other substances. It is stable under recommended storage conditions and should be stored in a tightly sealed container in a cool, well-ventilated area, protected from direct sunlight and ignition sources.
- Appearance: White to off-white solid
- Purity: 98.56% (LCMS)
- Molecular Formula: C28H40N4O6S
- Recommended storage for powder: -20°C for 3 years
- Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.