Learn More
Medchemexpress LLC (S,R,S)-AHPC-Boc-trans-3-aminocyclobutanol-Pip-CH2COOH | 2086301-47-3 | 99.3% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY131168100MG
Encompass_Preferred
(S,R,S)-AHPC-Boc-trans-3-aminocyclobutanol-Pip-CH2COOH is an E3 ligase ligand-linker conjugate. It incorporates a VHL ligand and is utilized in PROTAC technology. PROTACs are compounds that use the ubiquitin-proteasome system to degrade target proteins by linking an E3 ubiquitin ligase ligand with a target protein ligand. This product is for research use only, has a molecular weight of 754.98, a formula of C39H58N6O7S, and appears as a white to off-white solid.
  • E3 ligase ligand-linker conjugate
  • Incorporates a VHL ligand
  • Utilized in PROTAC technology
  • For research use only
  • White to off-white solid
Catalog No. 50-003-41081