Learn More
Medchemexpress LLC (S,R,S)-AHPC-Boc-trans-3-aminocyclobutanol-Pip-CH2COOH | 2086301-47-3 | 99.3% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY131168
Encompass_Preferred
(S,R,S)-AHPC-Boc-trans-3-aminocyclobutanol-Pip-CH2COOH is an E3 ligase ligand-linker conjugate that contains a VHL ligand on one end. It is used in PROTAC technology. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • E3 ligase ligand-linker conjugate
  • Contains a VHL ligand on one end
  • Used in PROTAC technology
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-42298