Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S,R,S)-AHPC-Boc-trans-3-aminocyclobutanol-Pip-CH2COOH | 2086301-47-3 | 99.3% | C39H58N6O7S | 10MG
SDP

Supplier:  Medchemexpress LLC HY13116810MG

Encompass_Preferred

(S,R,S)-AHPC-Boc-trans-3-aminocyclobutanol-Pip-CH2COOH is a Boc-protected VH032-derived VHL ligand-linker conjugate supplied as a high-purity research reagent for PROTAC and E3 ligase recruitment studies. It is intended for biochemical and cellular experiments and includes solubility and formulation guidance for in vitro and in vivo use.

  • Boc-protected VHL ligand-linker conjugate suitable for PROTAC research.
  • High purity (99.31%).
  • Molecular weight 754.98 g/mol; formula C39H58N6O7S.
  • Soluble in DMSO at up to 100 mg/mL with ultrasonic assistance.
  • In vivo formulation examples provided for common vehicles.
  • Available in multiple small-mass package sizes for research use.

Catalog No. 50-002-61265


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.