Learn More
Medchemexpress LLC (S,R,S)-AHPC-Boc-trans-3-aminocyclobutanol-Pip-CH2COOH | 2086301-47-3 | 99.31% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13116850MG
Encompass_Preferred
(S,R,S)-AHPC-Boc-trans-3-aminocyclobutanol-Pip-CH2COOH is an E3 ligase ligand-linker conjugate that incorporates a VHL ligand on one end. It is utilized in PROTAC (Proteolysis Targeting Chimeras) technology. PROTACs function by connecting a ligand for an E3 ubiquitin ligase with a ligand for a target protein via a linker, leveraging the intracellular ubiquitin-proteasome system to facilitate selective degradation of target proteins.
  • E3 ligase ligand-linker conjugate
  • Contains a VHL ligand
  • Used in PROTAC technology
  • Leverages the ubiquitin-proteasome system
  • Facilitates selective degradation of target proteins
Catalog No. 50-003-39883