Learn More
Medchemexpress LLC (S,R,S)-AHPC-Boc (VH032-Boc) | 1448189-98-7 | MFCD31692982 | 96.4% | 530.68 g·mol⁻¹ | C27H38N4O5S | 10MG
SDP
Supplier:  Medchemexpress LLC HY12310910MG
Encompass_Preferred
(S,R,S)-AHPC-Boc (VH032-Boc) is a small-molecule ligand that binds the von Hippel-Lindau (VHL) E3 ligase and is used as a VHL-recruiting moiety in PROTAC-mediated targeted protein degradation. It is supplied as solid material in multiple package sizes and as a 10 mM solution in DMSO. Molecular formula C27H38N4O5S, molecular weight 530.68 g·mol⁻¹, typical purity 96.4%.
  • Functions as a VHL ligand for recruitment of E3 ligase.
  • Suitable for PROTAC synthesis and chemical biology studies.
  • High purity (96.4%) for research use.
  • Available as solid in multiple quantities and as a 10 mM solution in DMSO.
  • Molecular formula C27H38N4O5S and molecular weight 530.68 g·mol⁻¹.
Catalog No. 50-002-38390