Learn More
Medchemexpress LLC (S,R,S)-AHPC-C10-NH2 dihydrochloride | 2341796-75-4 | 96.7% | 686.78 g·mol⁻¹ | C33H53Cl2N5O4S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY129941A5MG
Encompass_Preferred
(S,R,S)-AHPC-C10-NH2 dihydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates a VHL ligand and a ten-carbon alkyl linker for use in PROTAC synthesis and chemical biology applications. It is supplied as the dihydrochloride salt and is characterized by formula C33H53Cl2N5O4S and a molecular weight of 686.78 g·mol⁻¹.
  • Designed as an E3 ligase ligand-linker conjugate for PROTAC development.
  • Provides a ten-carbon alkyl linker terminating in a primary amine for conjugation.
  • Supplied as a dihydrochloride salt for improved stability and handling.
  • Available in small research quantities suitable for assay development.
  • Characterized with specified purity for chemical biology use.
Catalog No. 50-002-63074