Learn More
Medchemexpress LLC (S,R,S)-AHPC-C2-amide-benzofuranylmethyl-pyridine | 2347517-69-3 | 98.6% | 750.91 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14702550MG
Encompass_Preferred
(S,R,S)-AHPC-C2-amide-benzofuranylmethyl-pyridine is a dual target PROTAC that can target the ubiquitination and degradation of Smad3, and also improve the HIF-α protein level. It possesses a multi-path anti-fibrosis function and a renal protection function for research of renal anemia. This compound can be utilized for research on prostate cancer and other cancers.
  • Targets ubiquitination and degradation of Smad3
  • Improves HIF-α protein level
  • Possesses multi-path anti-fibrosis function
  • Provides renal protection function for research of renal anemia
  • Degrades Smad3 protein in a concentration-dependent manner
  • Significantly upregulates HIF-2α protein level in rat renal fibroblasts
  • Can degrade Smad3 protein levels and downregulate the androgen receptor (AR) pathway in CWR22Rv1 cells
  • Suitable for research on prostate cancer and other cancers
Catalog No. 50-003-62405