Learn More
Medchemexpress LLC HY-114176 100mg Medchemexpress, (S,R,S)-AHPC-C3-NH2 CAS:2245697-83-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY114176100MG
Encompass_Preferred
Medchemexpress, HY-114176 100mg (S,R,S)-AHPC-C3-NH2 CAS:2245697-83-8 (S,R,S)-AHPC-C3-NH2 is a synthesized E3 ligase ligand-linker conjugate that incorporates the (S,R,S)-AHPC based VHL ligand and a linker used in PROTAC technology. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2468