Learn More
Medchemexpress LLC (S,R,S)-AHPC-(C3-PEG)2-C6-Cl | 1835705-61-7 | MFCD31560481 | 95.0% | 749.44 g/mol | C39H61ClN4O6S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10360850MG
Encompass_Preferred
(S,R,S)-AHPC-(C3-PEG)2-C6-Cl is a VHL ligand-linker conjugate intended for use in proteolysis-targeting chimera (PROTAC) research. It couples an (S,R,S)-AHPC VHL-binding moiety to a short PEG linker and a chlorinated C6 terminus to enable E3 ligase recruitment and target degradation in cellular assays.
  • VHL ligand-linker conjugate suitable for PROTAC applications.
  • Demonstrated to induce degradation of GFP-HaloTag7 in cell-based assays.
  • High solubility in DMSO and ethanol; in vivo formulation options available.
  • Stored at -80°C, protected from light, under inert atmosphere.
  • Molecular weight 749.44 g/mol; chemical formula C39H61ClN4O6S.
Catalog No. 50-002-44605