Learn More
Medchemexpress LLC S R S-AHPC-C4-NH2 hy 2g | 2245697-83-8 | 2 G
SDP
Supplier:  Medchemexpress LLC HY1141762G
Encompass_Preferred
(S,R,S)-AHPC-C4-NH2 hydrochloride is an E3 ligase VHL ligand-linker conjugate supplied as a hydrochloride salt and used as a building block for PROTAC synthesis, particularly for VHL-based and EED-targeted degraders. It is provided in solid form or as ready-to-use DMSO solutions to support conjugation, screening, and medicinal chemistry workflows.
  • E3 ligase VHL ligand-linker conjugate suitable for PROTAC design.
  • Hydrochloride salt supplied as a solid with light yellow to green yellow color.
  • Available as solid and as 10 mM DMSO solution for ready reconstitution.
  • Intended as a synthetic building block for degrader molecules.
  • Documented purity and supporting datasheet and SDS available.
Catalog No. 50-003-07553