Learn More
Medchemexpress LLC HY-103605 100mg Medchemexpress, (S,R,S)-AHPC-C6-PEG3-C4-Cl CAS:1835705-55-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY103605100MG
Encompass_Preferred
Medchemexpress, HY-103605 100mg (S,R,S)-AHPC-C6-PEG3-C4-Cl CAS:1835705-55-9 (S,R,S)-AHPC-C6-PEG3-C4-Cl is a small molecule HaloPROTAC that incorporates the (S,R,S)-AHPC based VHL ligand and 3-unit PEG linker. (S,R,S)-AHPC-C6-PEG3-C4-Cl is capable of inducing the degradation of GFP-HaloTag7 in cell-based assays. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8140